Learn More
Saflufenacil-d7, TRC
Product Code. 30417560
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417560 0.5 mg 0.5mg
30424614 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30417560 Supplier Toronto Research Chemicals Supplier No. TRCS0818020.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2469260-39-5
Molecular Formula C17H10D7ClF4N4O5S
Molecular Weight (g/mol) 507.9
Synonym 2-Chloro-5-[3,6-dihydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)-1(2H)-pyrimidinyl]-4-fluoro-N-[[methyl(1-methylethyl)amino]sulfonyl]benzamide-d7, 2-Chloro-4-fluoro-N-[isopropyl(methyl)sulfamoyl]-5-[3-methyl-2,6-dioxo-4-(trifluoromethyl)pyrimidin-1-yl]benzamide-d7, 2-Chloro-5-[3,6-dihydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)-1(2H)-pyrimidinyl]-4-fluoro-N-[(isopropyl)methylsulfamoyl]benzamide-d7, 2-Chloro-5-[3,6-dihydro-3-methyl-2,6-dioxo-4-trifluoromethyl-1(2H)-pyrimidinyl]-4-fluoro-N-[[N-methyl-N-(1-methylethyl)amino]sulfonyl]benzamide-d7, Benmihuangcaoan-d7, Eragon-d7, Kixor-d7, N'-[2-Chloro-4-fluoro-5-[1,2,3,6-tetrahydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)pyrimidin-1-yl]benzoyl]-N-isopropyl-N-methylsulfamide-d7, Sharpen-d7, Treevix-d7
IUPAC Name 2-chloro-4-fluoro-5-(3-methyl-2,6-dioxo-4-(trifluoromethyl)-3,6-dihydropyrimidin-1(2H)-yl)-N-(N-methyl-N-(propan-2-yl-d7)sulfamoyl)benzamide
SMILES O=C(NS(=O)(N(C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C)=O)C1=CC(N(C(N(C)C(C(F)(F)F)=C2)=O)C2=O)=C(F)C=C1Cl
Specifications
Quantity 0.5 mg
InChI Formula InChI=1S/C17H17ClF4N4O5S/c1-8(2)25(4)32(30,31)23-15(28)9-5-12(11(19)6-10(9)18)26-14(27)7-13(17(20,21)22)24(3)16(26)29/h5-8H,1-4H3,(H,23,28)/i1D3,2D3,8D
Formula Weight 507.1
Chemical Name or Material Saflufenacil-d7