Description
- Disposable hygiene labcoats protect the work environment from particles coming from the operator's body (skin flakes, hair, sweat, etc.)
- They are suitable for HACCP procedures
- They help the operator protect themselves from the risk of stains related to their activity
Specifications
Specifications
| Size | Medium |
| Gender | Unisex |
| Material | Polypropylene |
| Color | White |
| Closure Type | Press Studs |
| Cuff Style | Elastic |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Resistance | None |
| Style | Knee-length |
| Show More |