Learn More
SymLine™ Signalbox and Switch Amplifier Accessories
Product Code. 10669290
Change view
Click to view available options
Includes:
10 Dual Lock, Reclosable Fastening Systems
Cable Set
Cable for Relay Contact
Disc Sensor, Alarm at Full State
Disc Sensor, Empty Level
EU Power Supply (220/230 V)
Holder for disc sensor
Hook and Loop Fastener Tape
Housing for Isolating Switch Amplifier
Signal Cable (10 m)
Signal Cable (3 m)
Signal Cable (5 m)
Signal Lamp, Magnetic Holder, Cable (3 m)
Product Type:
Disc Sensor
Dual Lock Closure
Holder
Housing
Power Supply
Signal Box Display
Signal Cable
Unit Size:
1 set
1 sheet
Each
Pack of 10
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Product Type Unit Size
10669290 Signal Cable (3 m) Signal Cable Each
10104574 Signal Cable (5 m) Signal Cable Each
10202570 Signal Cable (10 m) Signal Cable Each
10810571 Disc Sensor, Empty Level Disc Sensor Each
10337759 Disc Sensor, Alarm at Full State Disc Sensor Each
15912531 Cable Set Signal Cable 1 set
10657145 Cable for Relay Contact Signal Cable Each
10367589 EU Power Supply (220/230 V) Power Supply Each
16593532 Housing for Isolating Switch Amplifier Housing Each
10810021 10 Dual Lock, Reclosable Fastening Systems Dual Lock Closure Pack of 10
10121310 Hook and Loop Fastener Tape Dual Lock Closure Each
18676478 Holder for disc sensor Holder 1 sheet
18606488 Signal Lamp, Magnetic Holder, Cable (3 m) Signal Box Display 1 sheet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 10669290 Supplier SymLine™ Supplier No. 108050
  • ONLINE EXCLUSIVE PRICE
    49.15 EUR
    45.57 EUR / Pack of 1
    Save 3.58 EUR
    7% Off
Estimated Shipment: 03-12-2026
Accessories for the Built-in Signalbox and the Switch Amplifier.
  • LEDs and acoustic buzzer warn you in due time before the filling level of your collection containers reaches a critical height
  • When container is changed, the alert can be muted via the new touchpad
  • Pumps and other external devices can be conveniently triggered via the integrated interface
  • According to ATEX Directives, Ex-sensors have to be installed in conjunction with a switch amplifier when used in explosion-prone areas (Ex-zones)
  • Signalboxes and switch amplifier must be placed outside of the Ex-zone
  • Monitor two filling levels at once
  • Touchpad
  • Level control through status LEDs
  • Optical and acoustic alarm signals
  • Compatible with Switch AmplifierATEX: A widely used synonym for the ATEX directives of the European Union. The term ATEX is derived from the French abbreviation for ATmosphere EXplosive. Currently, this directive comprises two EU Directives on explosion prevention, namely the ATEX Equipment Directive 2014/34/EU and the ATEX Workplace Directive 1999/92/EC.
Specifications
Length (Metric) 3 m
Includes Signal Cable (3 m)
For Use With (Application) Accessorizing Signalbox and Switch Amplifier
For Use With (Equipment) Electronic Level Control
Product Type Signal Cable