Learn More
Deutsch Neumann™ Safety Gas Hoses
Product Code. 10077760
Change view
Click to view available options
Length (Metric):
500 mm
600 mm
750 mm
1000 mm
1250 mm
Unit Size:
1250mm
1500mm
500mm
600mm
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10077760 500 mm 500mm
10119510 600 mm 600mm
10684271 750 mm Each
10383281 1000 mm 1250mm
10129700 1250 mm 1500mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10077760 Supplier Deutsch Neumann™ Supplier No. 3608050
Provides safe and easy connection to gas burners. Deutsch Neumann™ Safety Gas Hoses consist of a rubber tube, threaded metal spiral and end sockets on both sides.
  • Working pressure to 100 mbar
  • For gas burners meeting DIN 30 665
  • For all gases meeting DVGW Work Code G 260
Specifications
Product Type Gas Hose
For Use With (Equipment) Gas burners meeting DIN 30 665 and all gases
Length (Metric) 500 mm
For Use With (Application) All Gases Meeting DVGW Work Code G 260