Learn More
Heidolph™ Safety Cover for Heat-On Block
Product Code. 13400049 Shop All Heidolph Instruments Products
Change view
Click to view available options
For Use With (Equipment):
1 L Round Bottom Flask
200 to 300 mL Round Bottom Flask
500 mL Round Bottom Flask
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
13400049 200 to 300 mL Round Bottom Flask Each
12397353 500 mL Round Bottom Flask Each
11367080 1 L Round Bottom Flask Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13400049 Supplier Heidolph™ Supplier No. 5058008100
  • ONLINE EXCLUSIVE PRICE
    532.00 EUR
    517.00 EUR / Pack of 1
    Save 15.00 EUR
    2% Off
Estimated Shipment: 30-11-2026
This cover for Heat-On Blocks provides an extra level of protection against burns when heating round bottom flasks.
  • Ensures more protection against burns when using Heat-On Block
  • Despite reduced danger when using Heat-On Blocks as compared to conventional heating bath, risk of burns is not zero- this safety cover reduces that
  • Cover is made of chemical-resistant, easy-to-clean, PTFE
TRUSTED_SUSTAINABILITY
Specifications
Color White
For Use With (Equipment) 200 to 300 mL Round Bottom Flask
For Use With (Application) Heat Protection
Material PTFE