Learn More
BOLA Safe-Lab Nuts
Product Code. 10471302
Change view
Click to view available options
Product Line:
NS 19/26
NS 29/32
NS 45/40
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Line Unit Size
10471302 NS 29/32 Each
10690942 NS 19/26 Each
10216981 NS 45/40 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10471302 Supplier BOLA Supplier No. K 134910
  • ONLINE EXCLUSIVE PRICE
    24.60 EUR
    18.35 EUR / Pack of 1
    Save 6.25 EUR
    25% Off
Estimated Shipment: 20-01-2027
Safe-Lab Nuts glass-fibre reinforced PTFE
  • Allows a tight and safe connection as well as an easy disconnection of cone and socket.
  • The special nut which is held on an external thread above the cone holds and locks the socket. For disconnection, this special nut has to be turned clockwise.
  • The power is enforced by the thread pitch and is transferred axially to the socket.
  • The ground joint is released.
Specifications
Certifications/Compliance FDA
Autoclavable Autoclavable
Product Line NS 29/32
Color White
Flexibility Rigid
Max. Operating Temperature (English) 482°F
Max. Operating Temperature (Metric) 250°C