missing translation for 'onlineSavingsMsg'
Learn More

Saccharin, Sigma-Aldrich™

Product Code. 15810771
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15810771 1 kg 1kg
15800771 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15810771 Supplier Sigma Aldrich Supplier No. 8201281000
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-07-2

Specifications

Melting Point 228°C to 230°C
MDL Number MFCD00005866
Quantity 1 kg
Synonym Saccharin, Saccharin
Molecular Weight (g/mol) 183.18
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.