missing translation for 'onlineSavingsMsg'
Learn More

Scharlab Sabouraud Dextrose Agar

Product Code. 11401867
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11401867 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11401867 Supplier Scharlab Supplier No. 01165500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Scharlab Sabouraud Dextrose Agar is for the enumeration and cultivation of fungi.

  • BAM/EP/HP/ISO/USP
  • Solid medium for the enumeration and cultivation of fungi
  • According to the Pharmacopeial Harmonised Method and ISO 13681 standard.
  • This medium need to add Chloramphenicol Selective Supplement (Art. No. 06-118-LYO1) and Oxytetracycline Selective Supplement (Art. No. 06-115LYO1).

Specifications

Certifications/Compliance ISO 13681
Form Solid
Media Type Sabouraud Dextrose Agar
Product Type Agar
Formulation D(+)-Glucose 40.00 g/L, Meat Peptone 5.00 g/L, Casein Peptone 5.00 g/L, Agar 15.00 g/L
For Use With (Application) Enumeration and Cultivation of Fungi
Incubation Conditions 20°C to 25°C
Incubation Time ≤ 5 Days
pH 5.6 ±0.2
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.