Learn More
S119-8, MedChemExpress
Product Code. 16493176
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493176 25 mg 25mg
16470937 50 mg 50mg
16444648 10 mg 10mg
16417367 5 mg 5mg
16415486 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16493176 Supplier Medchem Express Supplier No. HY112543
MedChemExpress S119-8 is a broad spectrum inhibitor of influenza A and B viruses, showing activity against multiple influenza B viruses and an oseltamivir-resistant influenza A virus, but does not inhibit a non-influenza virus, vesicular stomatitis nirus (VSV).
S119-8 is a broad spectrum inhibitor of influenza A and B viruses, showing activity against multiple influenza B viruses and an oseltamivir-resistant influenza A virus, but does not inhibit a non-influenza virus, vesicular stomatitis nirus (VSV).
Specifications
Chemical Name or Material S119-8
Physical Form Solid
CAS 443639-96-1
Color Light Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C23H24N2O
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : ≥ 150 mg/mL (435.48 mM)
SMILES O=C(NC1=CC=C(NC2=CC=CC=C2)C=C1)C3=CC=C(C(C)(C)C)C=C3
Molecular Weight (g/mol) 344.45
Formula Weight 344.45
Percent Purity 99.67%
Grade Research