Learn More
(S)-Xanthoanthrafil, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-Xanthoanthrafil |
| CAS | 247567-47-1 |
| Molecular Formula | C19 H23 N3 O6 |
| Quantity | 1 mg |
| Synonym | N-[(3,4-Dimethoxyphenyl)methyl]-2-[[(1S)-2-hydroxy-1-methylethyl]amino]-5-nitro-benzamide |
| InChI Formula | InChI=1S/C19H23N3O6/c1-12(11-23)21-16-6-5-14(22(25)26)9-15(16)19(24)20-10-13-4-7-17(27-2)18(8-13)28-3/h4-9,12,21,23H,10-11H2,1-3H3,(H,20,24)/t12-/m0/s1 |
| SMILES | COc1ccc(CNC(=O)c2cc(ccc2N[C@@H](C)CO)[N+](=O)[O-])cc1OC |
| IUPAC Name | N-[(3,4-dimethoxyphenyl)methyl]-2-[[(2S)-1-hydroxypropan-2-yl]amino]-5-nitrobenzamide |
| Molecular Weight (g/mol) | 389.402 |
| Formula Weight | 389.159 |