Learn More
(S)-Vigabatrin, TRC
Product Code. 30456761
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456761 2.5 mg 2.50mg
30456475 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30456761 Supplier Toronto Research Chemicals Supplier No. TRCV2530002.5MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 74046-07-4
Molecular Formula C6 H11 N O2
Molecular Weight (g/mol) 129.16
Synonym S-Vigabatrin
IUPAC Name (4S)-4-aminohex-5-enoic acid
SMILES N[C@@H](CCC(=O)O)C=C
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C6H11NO2/c1-2-5(7)3-4-6(8)9/h2,5H,1,3-4,7H2,(H,8,9)/t5-/m1/s1
Formula Weight 129.079
Chemical Name or Material (S)-Vigabatrin