missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Serine, Sigma-Aldrich™

Product Code. 15860221
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15860221 100 g 100g
15850221 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15860221 Supplier Sigma-Aldrich Supplier No. 8160200100
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

(S)-(+)-Serine is a non-essential amino acid that serves as a building block in peptide synthesis and as a precursor for various biomolecules, including neurotransmitters and phospholipids.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-45-1
Grade Research
Molecular Weight (g/mol) 105.09

Specifications

Melting Point 215°C to 225°C (Decomposes)
MDL Number MFCD00064224
Synonym (S)-(+)-Serine, 3-Hydroxy-L-alanine, Ser, (S)-(+)-Serine
Quantity 100 g
Chemical Name or Material (S)-(+)-Serine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.