Learn More
(S,S)-(+)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(ii), TRC
Product Code. 30454851
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454851 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30454851 Supplier Toronto Research Chemicals Supplier No. TRCT8888452.5G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (S,S)-(+)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(ii)
CAS 188264-84-8
Molecular Formula C36H52CoN2O2
Quantity 2.5 g
Synonym (SP-4-2)-[[2,2'-[(1S,2S)-1,2-Cyclohexanediylbis[(nitrilo-kN)methylidyne]]bis[4,6-bis(1,1-dimethylethyl)phenolato-kO]](2-)]cobalt, (S,S)-Jacobsen HKR catalyst
InChI Formula InChI=1S/C36H54N2O2.Co/c1-33(2,3)25-17-23(31(39)27(19-25)35(7,8)9)21-37-29-15-13-14-16-30(29)38-22-24-18-26(34(4,5)6)20-28(32(24)40)36(10,11)12;/h17-22,29-30,39-40H,13-16H2,1-12H3;/q;+2/p-2/t29-,30-;/m0./s1
SMILES CC(C)(C)c1cc2C=[N]3[C@H]4CCCC[C@@H]4[N]5=Cc6cc(cc(c6O[Co]35Oc2c(c1)C(C)(C)C)C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 603.74
Formula Weight 603.3361