Learn More
MedChemExpress (S,R,S)-AHPC-PEG4-N3
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
Specifications
Specifications
| Molecular Weight (g/mol) | 689.82 g/mol |
| CAS | 1797406-81-5 |
| Content And Storage | Pure form -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Quantity | 500 mg |
| Synonym | VH032-PEG4-N3 ∣ VHL Ligand-Linker Conjugates 5 ∣ E3 ligase Ligand-Linker Conjugates 4 |
| Solubility | DMSO : 50 mg/mL (72.48 mM; Need ultrasonic) |