Learn More
MedChemExpress (S,R,S)-AHPC-PEG3-N3
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
Specifications
Specifications
| Molecular Weight (g/mol) | 645.77 g/mol |
| CAS | 1797406-80-4 |
| Content And Storage | Pure form -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Quantity | 500 mg |
| Synonym | VH032-PEG3-N3 ∣ VHL Ligand-Linker Conjugates 8 ∣ E3 ligase Ligand-Linker Conjugates 12 |
| Solubility | Ethanol : 100 mg/mL (154.85 mM; Need ultrasonic) ∣ DMSO : 50 mg/mL (77.43 mM; Need ultrasonic) |