Learn More
MedChemExpress (S,R,S)-AHPC-C6-PEG3-C4-Cl
Product Code. 16480096
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
Product Code. 16480096
Brand: MedChemExpress HY103605
MedChemExpress (S,R,S)-AHPC-C6-PEG3-C4-Cl (VH032-C6-PEG3-C4-Cl) is a conjugate of ligands for E3 and 20-atom-length linker.
The connector of linker is Halogen group. (S,R,S)-AHPC-C6-PEG3-C4-Cl incorporates the (S,R,S)-AHPC based VHL ligand and an alkyl/ether-based linker. (S,R,S)-AHPC-C6-PEG3-C4-Cl is capable of inducing the degradation of GFP-HaloTag7 in cell-based assays.
Specifications
Molecular Weight (g/mol) 751.42 g/mol
CAS 1835705-55-9
Content And Storage Pure form -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Quantity 2 g
Synonym VH032-C6-PEG3-C4-Cl ∣ VHL Ligand-Linker Conjugates 12 ∣ E3 ligase Ligand-Linker Conjugates 8
Solubility DMSO : ≥ 100 mg/mL (133.08 mM) ∣ Ethanol : 100 mg/mL (133.08 mM; Need ultrasonic)