missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-Deacetyl Colchicine, TRC

Product Code. 30366614
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366614 25 mg 25mg
30363957 50 mg 50mg
30364864 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366614 Supplier Toronto Research Chemicals Supplier No. TRCD19892025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3476-50-4
Molecular Formula C20 H23 N O5
Molecular Weight (g/mol) 357.4
Synonym Benzo[a]heptalen-9(5H)-one, 7-amino-6,7-dihydro-1,2,3,10-tetramethoxy-, (7S)-, Benzo[a]heptalen-9(5H)-one, 7-amino-6,7-dihydro-1,2,3,10-tetramethoxy-, (S)-, Colchicine, N-deacetyl- (7CI,8CI), Colchicine, deacetyl- (6CI), (7S)-7-Amino-6,7-dihydro-1,2,3,10-tetramethoxybenzo[a]heptalen-9(5H)-one, (-)-7-Deacetylcolchicine, (-)-Deacetylcolchicine, Deacetylcolchicine, Desacetylcolchicine, N-Deacetylcolchicine, N-Desacetylcolchicine, NSC 201400
IUPAC Name (7S)-7-amino-1,2,3,10-tetramethoxy-6,7-dihydro-5H-benzo[a]heptalen-9-one
SMILES COC1=CC=C2C(=CC1=O)[C@@H](N)CCc3cc(OC)c(OC)c(OC)c23

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C20H23NO5/c1-23-16-8-6-12-13(10-15(16)22)14(21)7-5-11-9-17(24-2)19(25-3)20(26-4)18(11)12/h6,8-10,14H,5,7,21H2,1-4H3/t14-/m0/s1
Formula Weight 357.1576
Percent Purity >95%
Chemical Name or Material N-Deacetylcolchicine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.