missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-Deacetyl Colchicine, TRC

Product Code. 30363957
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363957 50 mg 50mg
30366614 25 mg 25mg
30364864 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363957 Supplier Toronto Research Chemicals Supplier No. TRCD19892050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3476-50-4
Molecular Formula C20 H23 N O5
Molecular Weight (g/mol) 357.4
Synonym Benzo[a]heptalen-9(5H)-one, 7-amino-6,7-dihydro-1,2,3,10-tetramethoxy-, (7S)-, Benzo[a]heptalen-9(5H)-one, 7-amino-6,7-dihydro-1,2,3,10-tetramethoxy-, (S)-, Colchicine, N-deacetyl- (7CI,8CI), Colchicine, deacetyl- (6CI), (7S)-7-Amino-6,7-dihydro-1,2,3,10-tetramethoxybenzo[a]heptalen-9(5H)-one, (-)-7-Deacetylcolchicine, (-)-Deacetylcolchicine, Deacetylcolchicine, Desacetylcolchicine, N-Deacetylcolchicine, N-Desacetylcolchicine, NSC 201400
IUPAC Name (7S)-7-amino-1,2,3,10-tetramethoxy-6,7-dihydro-5H-benzo[a]heptalen-9-one
SMILES COC1=CC=C2C(=CC1=O)[C@@H](N)CCc3cc(OC)c(OC)c(OC)c23

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C20H23NO5/c1-23-16-8-6-12-13(10-15(16)22)14(21)7-5-11-9-17(24-2)19(25-3)20(26-4)18(11)12/h6,8-10,14H,5,7,21H2,1-4H3/t14-/m0/s1
Formula Weight 357.1576
Percent Purity >95%
Chemical Name or Material N-Deacetylcolchicine
Grade HPLC
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.