missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-N-Benzyl-1-phenylethylamine, ChiPros∣r, 99%, ee 99+%

Product Code. p-7047476
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394186 5 g 5g
11304196 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394186 Supplier Thermo Scientific Alfa Aesar Supplier No. L19299.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17480-69-2 | C15H24N | 218.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17480-69-2
Molecular Formula C15H24N
Molecular Weight (g/mol) 218.36
MDL Number MFCD00066325
InChI Key UHABCGJJMSQRRP-ZDUSSCGKSA-O
Synonym s---n-benzyl-1-phenylethylamine, s---n-benzyl-alpha-methylbenzylamine, s-n-benzyl-1-phenylethanamine, s-n-benzyl-1-phenylethylamine, benzyl 1s-1-phenylethyl amine, s-n-benzyl-alpha-methylbenzylamine, s--n-benzyl-alpha-phenylethylamine, pubchem5973, n-benzyl-d-amphetamine
PubChem CID 1268085
IUPAC Name (1S)-N-benzyl-1-phenylethanamine
SMILES C[C@H]([NH2+]CC1CCCCC1)C1=CC=CC=C1

Specifications

Density 1.01
Boiling Point 171°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.564
Quantity 5 g
Beilstein 3650264
Sensitivity Air Sensitive
Optical Rotation −39° (Neat)
Formula Weight 211.31
Percent Purity 99%
Assay ee ≥99%
Chemical Name or Material (S)-(-)-N-Benzyl-1-phenylethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.