missing translation for 'onlineSavingsMsg'
Learn More

(S)-Ketorolac, MedChemExpress

Product Code. 16493786
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493786 25 mg 25mg
16481008 5 mg 5mg
16408538 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16493786 Supplier Medchem Express Supplier No. HYB0580A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S)-Ketorolac is a nonsteroidal anti-inflammatory agent. (S)-ketorolac exhibits potent COX1 and COX2 enzyme inhibition.

(S)-Ketorolac is a nonsteroidal anti-inflammatory agent. (S)-ketorolac exhibits potent COX1 and COX2 enzyme inhibition.

Specifications

Chemical Name or Material (S)-Ketorolac
CAS 66635-92-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H13NO3
Quantity 25 mg
Synonym (-)-Ketorolac
Solubility Information DMSO : 200 mg/mL (783.48 mM; Need ultrasonic)
SMILES O=C(C1=CC=CC=C1)C2=CC=C3N2CC[C@@H]3C(O)=O
Molecular Weight (g/mol) 255.27
Formula Weight 255.27
Percent Purity 99.62%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.