missing translation for 'onlineSavingsMsg'
Learn More

(S)-CPW 399, TRC

Product Code. 30364316
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364316 50 mg 50mg
30367889 100 mg 100mg
30359045 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364316 Supplier Toronto Research Chemicals Supplier No. TRCC78147850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 389888-02-2
Molecular Formula C10H13N3O4
Molecular Weight (g/mol) 239.23
Synonym (S)-CPW 399, (2S)-2-Amino-3-(2,4-dioxo-6,7-dihydro-5H-cyclopenta[d]pyrimidin-1-yl)propanoic Acid, (S)-2-Amino-3-(2,4-dioxo-2,3,4,5,6,7-hexahydro-1H-cyclopenta[d]pyrimidin-1-yl)propanoic Acid
IUPAC Name (2S)-2-amino-3-(2,4-dioxo-6,7-dihydro-5H-cyclopenta[d]pyrimidin-1-yl)propanoic acid
SMILES N[C@@H](CN1C(=O)NC(=O)C2=C1CCC2)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H13N3O4/c11-6(9(15)16)4-13-7-3-1-2-5(7)8(14)12-10(13)17/h6H,1-4,11H2,(H,15,16)(H,12,14,17)/t6-/m0/s1
Formula Weight 239.0906
Chemical Name or Material (alphaS)-alpha-Amino-2,3,4,5,6,7-hexahydro-2,4-dioxo-1H-cyclopentapyrimidine-1-propanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.