CAS RN 389888-02-2
IUPAC Name:
2-amino-3-{2,4-dioxo-1H,2H,3H,4H,5H,6H,7H-cyclopenta[d]pyrimidin-1-yl}propanoic acid
Synonyms:
2-amino-3-{2,4-dioxo-3H,5H,6H,7H-cyclopenta[d]pyrimidin-1-yl}propanoic acid
Molecular Weight (g/mol):
239.23
Molecular Formula:
C10H13N3O4
InChi Key:
VSGUEKZRMJVQOH-UHFFFAOYNA-N
SMILES:
NC(CN1C2=C(CCC2)C(=O)NC1=O)C(O)=O
1
–
1
of
1
results
1
(S)-CPW 399, TRC
CAS: 389888-02-2 Molecular Formula: C10H13N3O4 Molecular Weight (g/mol): 239.23 Synonym: (S)-CPW 399,(2S)-2-Amino-3-(2,4-dioxo-6,7-dihydro-5H-cyclopenta[d]pyrimidin-1-yl)propanoic Acid,(S)-2-Amino-3-(2,4-dioxo-2,3,4,5,6,7-hexahydro-1H-cyclopenta[d]pyrimidin-1-yl)propanoic Acid IUPAC Name: (2S)-2-amino-3-(2,4-dioxo-6,7-dihydro-5H-cyclopenta[d]pyrimidin-1-yl)propanoic acid SMILES: N[C@@H](CN1C(=O)NC(=O)C2=C1CCC2)C(=O)O