Learn More
(S)-(-)-Bay-K-8644, MedChemExpress
Product Code. 16476318
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476318 10 mg 10mg
16432437 50 mg 50mg
16428907 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16476318 Supplier Medchem Express Supplier No. HY15124
MedChemExpress (S)-(-)-Bay-K-8644 is an agonist of L-type Ca2+ channel. (S)-(-)-Bay-K-8644 activates Ba2+ currents (IBa) (EC50=32 nM).
(S)-(-)-Bay-K-8644 is an agonist of L-type Ca2+ channel. (S)-(-)-Bay-K-8644 activates Ba2+ currents (IBa) (EC50=32 nM).
Specifications
Chemical Name or Material (S)-(-)-Bay-K-8644
CAS 98625-26-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H15F3N2O4
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (350.83 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CC1=C([N+]([O-])=O)[C@@H](C2=C(C(F)(F)F)C=CC=C2)C(C(OC)=O)=C(C)N1
Molecular Weight (g/mol) 356.3
Formula Weight 356.3
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation