missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Arginine Hydrochloride, Sigma-Aldrich™

Product Code. 15850191
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15850191 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15850191 Supplier Sigma-Aldrich Supplier No. 8160010100
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

Melting Point >235°C
CAS 1119-34-2
MDL Number MFCD00064550
Synonym ʟ-Arginine monohydrochloride, α-Amino-δ-guanidino valerianic acid hydrochloride, alpha-Amino-delta-guanidino valerianic acid hydrochloride, Arg, (S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride, ʟ-Arginine monohydrochloride, (S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride
Density 1.42 g/cm3 at 20°C
Molecular Weight (g/mol) 210.66
Quantity 100 g
Grade Research
Chemical Name or Material (S)-(+)-Arginine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.