Learn More
(S)-7,8-Dihydroxy Efavirenz, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-7,8-Dihydroxy Efavirenz |
| CAS | 2454245-79-3 |
| Molecular Formula | C14H9ClF3NO4 |
| Quantity | 2.5 mg |
| Synonym | (S)-6-Chloro-4-(cyclopropylethynyl)-7,8-dihydroxy-4-(trifluoromethyl)-1H-benzo[d][1,3]oxazin-2(4H)-one |
| InChI Formula | InChI=1S/C14H9ClF3NO4/c15-8-5-7-9(11(21)10(8)20)19-12(22)23-13(7,14(16,17)18)4-3-6-1-2-6/h5-6,20-21H,1-2H2,(H,19,22)/t13-/m0/s1 |
| SMILES | Oc1c(O)c2NC(=O)O[C@@](C#CC3CC3)(c2cc1Cl)C(F)(F)F |
| Molecular Weight (g/mol) | 347.67 |
| Formula Weight | 347.0172 |