Learn More
(S)-4,4,7A-Trimethyl-5,6,7,7A-tetrahydrobenzofuran-2(4H)-one, TRC
Specifications
Specifications
| Chemical Name or Material | (7aS)-4,4,7a-Trimethyl-5,6,7,7a-tetrahydrobenzofuran-2(4H)-one |
| CAS | 81800-41-1 |
| Molecular Formula | C11 H16 O2 |
| Quantity | 1 g |
| Synonym | (7aS)-4,4,7a-Trimethyl-5,6,7,7a-tetrahydrobenzofuran-2(4H)-one, 2(4H)-Benzofuranone','5,6,7,7a-tetrahydro-4,4,7a-trimethyl-','(7aS)-, 2(4H)-Benzofuranone','5,6,7,7a-tetrahydro-4,4,7a-trimethyl-','(S)-, (7aS)-5,6,7,7a-Tetrahydro-4,4,7a-trimethyl-2(4H)-benzofur |
| InChI Formula | InChI=1S/C11H16O2/c1-10(2)5-4-6-11(3)8(10)7-9(12)13-11/h7H,4-6H2,1-3H3/t11-/m0/s1 |
| SMILES | CC1(C)CCC[C@]2(C)OC(=O)C=C12 |
| IUPAC Name | (7aS)-4,4,7a-trimethyl-6,7-dihydro-5H-1-benzofuran-2-one |
| Molecular Weight (g/mol) | 180.244 |
| Formula Weight | 180.115 |