Learn More
(S)-2-(Benzyloxy)propional, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-2-(Benzyloxy)propional |
| CAS | 81445-44-5 |
| Molecular Formula | C10 H12 O2 |
| Quantity | 50 mg |
| Synonym | Propanal, 2-(phenylmethoxy)-, (2S)-, Propanal, 2-(phenylmethoxy)-, (S)-, (2S)-2-(Phenylmethoxy)propanal, (-)-(S)-2-(Benzyloxy)propanal, (-)-(S)-2-(Phenylmethoxy)propanal, (2S)-2-(Benzyloxy)propanal, (2S)-2-Benzyloxypropanal, (2S)-2-Phenylmethoxypropanal, (S)-2-(Benzyloxy)propanal, (S)-2-(Benzyloxy)propionaldehyde, (S)-α-(Benzyloxy)propanal |
| InChI Formula | InChI=1S/C10H12O2/c1-9(7-11)12-8-10-5-3-2-4-6-10/h2-7,9H,8H2,1H3/t9-/m0/s1 |
| SMILES | C[C@H](OCc1ccccc1)C=O |
| IUPAC Name | (2S)-2-phenylmethoxypropanal |
| Molecular Weight (g/mol) | 164.2 |
| Formula Weight | 164.0837 |