Learn More
(S)-2-(Benzyloxy)propional, TRC
Product Code. 30343343
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30343343 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30343343 Supplier Toronto Research Chemicals Supplier No. TRCB287725250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (S)-2-(Benzyloxy)propional
CAS 81445-44-5
Molecular Formula C10 H12 O2
Quantity 250 mg
Synonym Propanal, 2-(phenylmethoxy)-, (2S)-, Propanal, 2-(phenylmethoxy)-, (S)-, (2S)-2-(Phenylmethoxy)propanal, (-)-(S)-2-(Benzyloxy)propanal, (-)-(S)-2-(Phenylmethoxy)propanal, (2S)-2-(Benzyloxy)propanal, (2S)-2-Benzyloxypropanal, (2S)-2-Phenylmethoxypropanal, (S)-2-(Benzyloxy)propanal, (S)-2-(Benzyloxy)propionaldehyde, (S)-α-(Benzyloxy)propanal
InChI Formula InChI=1S/C10H12O2/c1-9(7-11)12-8-10-5-3-2-4-6-10/h2-7,9H,8H2,1H3/t9-/m0/s1
SMILES C[C@H](OCc1ccccc1)C=O
IUPAC Name (2S)-2-phenylmethoxypropanal
Molecular Weight (g/mol) 164.2
Formula Weight 164.0837
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.