Learn More
S-2-Benzothiazolyl (Z)-2-(2-aminothiazole-4-yl)-2-methoxy-carbonylmethioacetate, TRC
Specifications
Specifications
| Chemical Name or Material | S-2-Benzothiazolyl (Z)-2-(2-aminothiazole-4-yl)-2-methoxy-carbonylmethioacetate |
| CAS | 246035-38-1 |
| Molecular Formula | C15H12N4O4S3 |
| Quantity | 100 mg |
| Synonym | Cefixime Impurity 2 |
| InChI Formula | InChI=1S/C15H12N4O4S3/c1-22-11(20)6-23-19-12(9-7-24-14(16)17-9)13(21)26-15-18-8-4-2-3-5-10(8)25-15/h2-5,7H,6H2,1H3,(H2,16,17)/b19-12- |
| SMILES | COC(=O)CO/N=C(/C1=CSC(=N1)N)\C(=O)SC2=NC3=CC=CC=C3S2 |
| IUPAC Name | methyl 2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-(1,3-benzothiazol-2-ylsulfanyl)-2-oxoethylidene]amino]oxyacetate |
| Molecular Weight (g/mol) | 408.47 |
| Formula Weight | 408.0021 |