missing translation for 'onlineSavingsMsg'
Learn More

S-(2-Aminoethyl)thiouronium Bromide Hydrobromide, Sigma-Aldrich™

Product Code. 15854481
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15854481 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15854481 Supplier Sigma Aldrich Supplier No. 8200640010
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 56-10-0
Melting Point 188°C to 191°C
Molecular Formula H2NCH2CH2SC(=NH)NH2 · 2HBr
MDL Number MFCD00037011
Quantity 10 g
Synonym S-(2-Aminoethyl)thiouronium bromide hydrobromide, AET, S-(2-Aminoethyl)thiouronium bromide hydrobromide
Molecular Weight (g/mol) 281.01
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.