Learn More
(S)-2-(3,4,5-Trimethoxyphenyl)butyric Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-2-(3,4,5-Trimethoxyphenyl)butyric Acid |
| CAS | 195202-08-5 |
| Molecular Formula | C13H18O5 |
| Quantity | 100 mg |
| Synonym | (alphaS)-alpha-Ethyl-3,4,5-trimethoxybenzeneacetic Acid |
| InChI Formula | InChI=1S/C13H18O5/c1-5-9(13(14)15)8-6-10(16-2)12(18-4)11(7-8)17-3/h6-7,9H,5H2,1-4H3,(H,14,15)/t9-/m0/s1 |
| SMILES | CC[C@H](C(=O)O)c1cc(OC)c(OC)c(OC)c1 |
| IUPAC Name | (2S)-2-(3,4,5-trimethoxyphenyl)butanoic acid |
| Molecular Weight (g/mol) | 254.28 |
| Formula Weight | 254.1154 |