Learn More
(S)-2-(3,4,5-Trimethoxyphenyl)butyric Acid, TRC
Product Code. 30455447
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455447 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455447 Supplier Toronto Research Chemicals Supplier No. TRCT797485250MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (S)-2-(3,4,5-Trimethoxyphenyl)butyric Acid
CAS 195202-08-5
Molecular Formula C13H18O5
Quantity 250 mg
Synonym (alphaS)-alpha-Ethyl-3,4,5-trimethoxybenzeneacetic Acid
InChI Formula InChI=1S/C13H18O5/c1-5-9(13(14)15)8-6-10(16-2)12(18-4)11(7-8)17-3/h6-7,9H,5H2,1-4H3,(H,14,15)/t9-/m0/s1
SMILES CC[C@H](C(=O)O)c1cc(OC)c(OC)c(OC)c1
IUPAC Name (2S)-2-(3,4,5-trimethoxyphenyl)butanoic acid
Molecular Weight (g/mol) 254.28
Formula Weight 254.1154