Learn More
(S)-(+)-1-Phenylethyl isothiocyanate, 97%, Thermo Scientific™
Product Code. 15479119
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15479119 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15479119 Supplier Thermo Scientific Alfa Aesar Supplier No. L19986.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
CAS 24277-43-8
Boiling Point 132°C to 133°C (20 mmHg)
Flash Point 103°C (217°F)
Odor Irritating
Molecular Formula C9H9NS
MDL Number MFCD02093279
Quantity 1 g
UN Number UN2922
Beilstein 6476424
Synonym 1s-1-isothiocyanatoethyl benzene, s-+-1-phenylethyl isothiocyanate, s-+-alpha-methylbenzyl isothiocyanate, l--methylbenzyl isothiocyanate, s-1-phenylethyl isothiocyanate, s---phenylethyl isothiocyanate, s-+-, a-methylbenzyl isothiocyanate, benzene, 1s-1-isothiocyanatoethyl, s + alpha-methylbenzyl isothiocyanate l, s-+-alpha-methylbenzyl isothiocyanate, for chiral derivatization
InChI Key QQCJPTVZIZVKEZ-QMMMGPOBSA-N
Optical Rotation +17° (c=10 in Chloroform)
SMILES CC(C1=CC=CC=C1)N=C=S
IUPAC Name [(1S)-1-isothiocyanatoethyl]benzene
Molecular Weight (g/mol) 163.238
PubChem CID 737164
Formula Weight 163.24
Percent Purity 97%
Chemical Name or Material (S)-(+)-1-Phenylethyl isothiocyanate