missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(Boc-amino)-2-propanol, 97%

Product Code. 15469597
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15469597 25 g 25g
15459597 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469597 Supplier Thermo Scientific Alfa Aesar Supplier No. H52826.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 167938-56-9 | C8H17NO3 | 175.228 g/mol

(S)-1-(Boc-amino)-2-propanol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-1-(Boc-amino)-2-propanol is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 167938-56-9
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.228
MDL Number MFCD04974340
InChI Key YNJCFDAODGKHAV-LURJTMIESA-N
Synonym n-boc-s-1-amino-2-propanol, s-tert-butyl 2-hydroxypropyl carbamate, s-1-boc-amino-2-propanol, tert-butyl n-2s-2-hydroxypropyl carbamate, tert-butyl 2s-2-hydroxypropyl carbamate, s-2-hydroxy-propyl-carbamic acid t-butyl ester, s-2-hydroxypropyl carbamic acid tert-butyl ester, s-2-hydroxy-propyl-carbamic acid tert-butyl ester
PubChem CID 11789722
IUPAC Name tert-butyl N-[(2S)-2-hydroxypropyl]carbamate
SMILES CC(CNC(=O)OC(C)(C)C)O

Spécification

Quantity 25 g
Formula Weight 175.23
Percent Purity 97%
Chemical Name or Material (S)-1-(Boc-amino)-2-propanol

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.