Learn More
(S)-1-Aminoindane, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-1-Aminoindane |
| CAS | 61341-86-4 |
| Molecular Formula | C9 H11 N |
| Quantity | 250 mg |
| Synonym | 1H-Inden-1-amine, 2,3-dihydro-, (1S)-, 1H-Inden-1-amine, 2,3-dihydro-, (S)-, (1S)-2,3-Dihydro-1H-inden-1-amine, ((1S)-Indan-1-yl)amine, (+)-1-Aminoindan, (1S)-1-Aminoindane, (1S)-2,3-Dihydro-1H-1-indenamine, (1S)-2,3-Dihydro-1H-inden-1-amine, (1S)-2,3-Dihydro-1H-inden-1-ylamine, (S)-(+)-1-Aminoindan, (S)-(+)-1-Aminoindane, (S)-(+)-Indan-1-ylamine, (S)-1-Aminoindan, (S)-2,3-Dihydro-1H-inden-1-amine, (S)-2,3-Dihydroinden-1-amine, 1(S)-Aminoindane, [(S)-Indan-1-yl]amine |
| InChI Formula | InChI=1S/C9H11N/c10-9-6-5-7-3-1-2-4-8(7)9/h1-4,9H,5-6,10H2/t9-/m0/s1 |
| SMILES | N[C@H]1CCc2ccccc12 |
| IUPAC Name | (1S)-2,3-dihydro-1H-inden-1-amine |
| Molecular Weight (g/mol) | 133.19 |
| Formula Weight | 133.0891 |