missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(2-Methoxyphenyl)ethylamine, ChiPros™, 99%, ee 98+%, Thermo Scientific™

Product Code. 11491540
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491540 5 g 5g
11481540 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491540 Supplier Thermo Scientific Alfa Aesar Supplier No. H26948.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(S)-1-(2-Methoxyphenyl)ethylamine play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks or chiral auxiliaries.

Solubility
Soluble in water(22 g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68285-24-5
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00671658
InChI Key VENQOHAPVLVQKV-ZETCQYMHSA-N
Synonym s-1-2-methoxyphenyl ethanamine, s-1-2-methoxyphenyl ethylamine, 1s-1-2-methoxyphenyl ethanamine, 1s-1-2-methoxyphenyl ethylamine, s-2-methoxy-methylbenzylamine, s-2-methoxy-, a-methylbenzylamine, s-2-methoxy-alpha-methylbenzylamine, 1s-1-2-methoxyphenyl-ethylamine, 1s-1-2-methoxyphenyl ethan-1-amine, s---2-methoxy a-methylbenzylamine hcl
PubChem CID 853082
IUPAC Name (1S)-1-(2-methoxyphenyl)ethanamine
SMILES CC(C1=CC=CC=C1OC)N

Specifications

Boiling Point 118°C (17 mmHg)
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Soluble in water(22g/L).
Formula Weight 151.21
Percent Purity 99%
Assay ee ≥98%
Chemical Name or Material (S)-1-(2-Methoxyphenyl)ethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.