Learn More
Fisherbrand™ Rulers
Product Code. 12979074 Shop All Fisherbrand Products
Change view
Click to view available options
Scale Type:
Metric
Metric and English
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Scale Type unitSize
12979074 Metric and English Pack of 10
12989074 Metric Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12979074
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
Moulded in white plastic.
  • 6mm thick by 25mm wide
  • 1 meter long
  • Foil printed in black
Specifications
Product Type Ruler
Color White
Length (Metric) 1 m
Scale Type Metric and English
Thickness 6 mm
Width (Metric) 25 mm
Material Plastic