Learn More
Brady™ Rugged Reinforced Nylon Safety Padlocks
Product Code. 15693557 Shop All Brady Products
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
White
Yellow
Width (Metric):
38 mm
Unit Size:
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Width (Metric) Unit Size
15693557 White 38 mm Pack of 6
15603557 Black 38 mm Pack of 6
15683547 Blue 38 mm Pack of 6
15673557 Brown 38 mm Pack of 6
15623557 Green 38 mm Pack of 6
15633557 Orange 38 mm Pack of 6
15683557 Purple 38 mm Pack of 6
15693547 Red 38 mm Pack of 6
15613557 Yellow 38 mm Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15693557 Supplier Brady™ Supplier No. 813641
  • ONLINE EXCLUSIVE PRICE
    138.00 EUR
    130.87 EUR / Pack of 6
    Save 7.13 EUR
    5% Off
In Stock
Brady™ Standard Safety Padlocks with Steel Shackle are the ultimate locks for all safety and security needs, and are precision-machined for better tamper resistance.
  • Unique cylinder is insulated to protect workers from shocks when key is inserted
  • Six-pin precision-machined for more unique key cuts and better tamper resistance
  • Reserved, paracentric keyway provides optimal security
  • Special design prohibits the key from being released until the padlock shackle is closed
  • Locks are compact and lightweight and resist impact, temperature, chemicals and corrosion
  • Non-conductive, non-sparking body features a ribbed design for easier gripping and handling
  • One key supplied with each lock
  • Key retaining - key will not release until the padlock shackle is securely closed
Specifications
Product Type Safety Padlocks
Key Type Keyed Different
Alternate Part Number SFTY PADLOCK NYLON 38MM SHA WHITE 6/PAC
Color White
Material Reinforced Nylon
Description Safety Padlocks - Nylon Shackle
Diameter (English) Shackle 0.25 in.
Diameter (Metric) Shackle 6.5 mm
Dimensions (L x W) 38.00 x 86.50 mm
For Use With (Application) Lockout Tagout
Height (Metric) 86.5 mm
Height (Metric) Shackle 38.00 mm
Material (Shackle) Nylon
No. of Pins 6
Quantity 6/Box
Width (Metric) 38 mm
Width (Metric) Shackle 33 mm