Learn More
Fisherbrand™ Rubber Stoppers With a Bore
Product Code. 11597592 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Outer:
6 mm
13 mm
35 mm
37 mm
41 mm
45 mm
53 mm
Unit Size:
Pack of 10
Pack of 2
Pack of 20
Pack of 5
Pack of 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer unitSize
11597592 13 mm Pack of 20
10283751 35 mm Pack of 10
11517612 37 mm Pack of 10
10000331 37 mm Pack of 10
11537612 41 mm Pack of 5
11557612 45 mm Pack of 5
10080361 53 mm Pack of 2
10440123 6 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11597592
  • ONLINE EXCLUSIVE PRICE
    7.35 EUR
    5.70 EUR / Pack of 20
    Save 1.65 EUR
    22% Off
View all promotions
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
Prevents liquid chemical leaks or escape from container.
  • Used for industrial purposes
  • Fits tightly to the container
  • Red natural rubber construction
Specifications
Quantity 20/Pk.
Color Red
Diameter (Metric) Outer 13 mm
Material Rubber
Product Type Stopper (1 hole)