Learn More
- Home
- Products
- Stoppers and Closures
- Corks
- Fisherbrand™ Cork Stoppers
Fisherbrand™ Cork Stoppers
Product Code. 12336467 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Bottom:
6 mm
8 mm
9 mm
12 mm
13 mm
14 mm
17 mm
19 mm
21 mm
22 mm
25.5 mm
28.5 mm
30 mm
32 mm
35 mm
38 mm
41.5 mm
44 mm
47.5 mm
50 mm
Diameter (Metric) Top:
9.5 mm
11 mm
12 mm
15 mm
16 mm
17 mm
20.5 mm
22 mm
24 mm
25.5 mm
28.5 mm
32 mm
33 mm
35 mm
38 mm
41.5 mm
44 mm
47.5 mm
50 mm
57 mm
Unit Size:
Pack of 10
Pack of 20
21 product options available for selection
| Product Code. | Diameter (Metric) Bottom | Diameter (Metric) Top | Unit Size |
|---|---|---|---|
| 12336467 | 25.5 mm | 28.5 mm | Pack of 10 |
| 12346457 | 12 mm | 15 mm | Pack of 20 |
| 12356457 | 13 mm | 16 mm | Pack of 20 |
| 12366457 | 14 mm | 17 mm | Pack of 20 |
| 12386457 | 17 mm | 20.5 mm | Pack of 10 |
| 12316467 | 21 mm | 24 mm | Pack of 10 |
| 12326467 | 22 mm | 25.5 mm | Pack of 10 |
| 12306467 | 19 mm | 22 mm | Pack of 10 |
| 12346467 | 28.5 mm | 32 mm | Pack of 10 |
| 12356467 | 30 mm | 33 mm | Pack of 10 |
| 12803834 | 30 mm | 33 mm | Pack of 10 |
| 12366467 | 32 mm | 35 mm | Pack of 10 |
| 12376467 | 35 mm | 38 mm | Pack of 10 |
| 12386467 | 38 mm | 41.5 mm | Pack of 10 |
| 12396467 | 41.5 mm | 44 mm | Pack of 10 |
| 12306477 | 44 mm | 47.5 mm | Pack of 10 |
| 12316477 | 47.5 mm | 50 mm | Pack of 10 |
| 12306457 | 6 mm | 9.5 mm | Pack of 20 |
| 12326477 | 50 mm | 57 mm | Pack of 10 |
| 12316457 | 8 mm | 11 mm | Pack of 20 |
| 12326457 | 9 mm | 12 mm | Pack of 20 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
Specifications
Specifications
| Length (Metric) | 32 mm |
| Quantity | 10/Pk. |
| Diameter (Metric) Bottom | 25.5 mm |
| Diameter (Metric) Top | 28.5 mm |
| Material | Cork |
| Product Type | Cork Stopper |