Learn More
Deutsch Neumann™ The Flip™ Safety Pipette Fillers
Click to view available options
:
Capacity: 0.1 to 100mL
Description
- Only two operating points
- Evacuate via the new automatic valve
- One-handed design is the simplest safety pipette filler available
- Ability to clean inside of bulb by removing patented valve and rinsing out
Specifications
Specifications
| Color | Red |
| Material | Rubber |
| Type | Flip Security Pear Pipette |
| Quantity | 1 Each |