Learn More
Abeba™ Rubber 9250 Shoes
Product Code. 15455713
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15455713 37 1 pair
15445713 36 1 pair
15465713 38 1 pair
15475713 39 1 pair
15485713 40 1 pair
15495713 41 1 pair
15415723 42 1 pair
15425723 43 1 pair
15435723 44 1 pair
15673208 45 1 pair
15683208 46 1 pair
15693208 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15455713 Supplier Abeba™ Supplier No. 9250/37
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 09-12-2026
Acid and base resistant. Rubber sole provides high slip resistance.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Dark blue
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC/li
Color Blue
Material Leather
Size 37
Sole Features Slip-resistant
Sole Material Rubber
Style 9250