Learn More
Abeba™ Rubber 5200 Shoes
Product Code. 15356691
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15356691 42 1 pair
15631838 36 1 pair
15396681 37 1 pair
15306691 38 1 pair
15641838 39 1 pair
15326691 40 1 pair
15346691 41 1 pair
15386691 43 1 pair
15346701 44 1 pair
15382335 45 1 pair
15392335 46 1 pair
15302345 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15356691 Supplier Abeba™ Supplier No. 5200/42
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 08-12-2026
ESD dissipative shoe is acid and base resistant.
  • CE EN ISO 20347:2012 OB SRC
  • Smooth leather white
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Leather top layer
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB SRC
Color White
Material Leather
Size 42
Sole Features Slip-resistant
Sole Material Rubber
Style 5200