Learn More
Jenway™ RS232 to USB Converter
Product Code. 11430609
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430609 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11430609 Supplier Jenway™ Supplier No. 9995979
  • ONLINE EXCLUSIVE PRICE
    86.00 EUR
    79.70 EUR / Pack of 1
    Save 6.30 EUR
    7% Off
Estimated Shipment: 28-12-2026
Converts RS232 to USB for computers without a serial port
  • Converts RS232 to USB for computers without a serial port
  • For use with Jenway Spectrophotmeters, Conductivity and pH Meters
Specifications
Quantity 1 Ea.
Product Type RS232 to USB Converter