Learn More
Thermo Scientific™ Round Bucket for Thermo Scientific™ Sorvall™ Stratos Rotor
Product Code. 10488732 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
18 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
10488732 18 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10488732 Supplier Thermo Scientific™ Supplier No. 75008172
  • ONLINE EXCLUSIVE PRICE
    421.00 EUR
    402.00 EUR / Pack of 1
    Save 19.00 EUR
    4% Off
In Stock
For use with Four-Place Swinging Bucket Rotor 75-003-047
Order Info
Order correct quantity to equip rotor. Aerosol-tight lids (75008173) sold separately. Optional adapters accept assorted tube styles and volumes from 1.5mL to 100mL.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Swing out rotor for Labofuge 400 and 400R centrifuge
Capacity 18 mL
Tube Size 1.5 to 100 mL