missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Rottlerin
GREENER_CHOICE

Product Code. p-4785451
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001271

Brand: Tocris Bioscience™ 1610/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reported PKCδ inhibitor

Originally reported to inhibit PKC isoforms. Also reported to inhibit CAM kinase III. However, recently shown to inhibit a wide range of protein kinases, and most potently to inhibit PRAK and MAPKAP-K2 (IC50 values are 1.9 and 5 μM respectively). Also shown to act as a direct mitochondrial uncoupler. Thought to stimulate autophagy by targeting upstream mTORC1 control pathways.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82-08-6
Molecular Formula C30H28O8

Specifications

Quantity 10 mg
Formula Weight 516.55
Purity >98%
Molecular Weight (g/mol) 516.546
Chemical Name or Material Rottlerin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.