Learn More
Eppendorf™ Rotor for Centrifuges 5910 Ri and 5920 R
Product Code. 15863212 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
20 × 5 mL
4 x 1,000 mL
4 x 750 mL
48 x 2 mL
6 x 250 mL
6 × 50 mL
Max. Speed:
10,100 rpm
13,100 rpm
13,500 rpm
13,700 rpm
4,500 rpm
4,700 rpm
Max. RCF:
15,054 x g
20,130 x g
20,913 x g
21,194 x g
4,347 x g
4,800 x g
Includes:
Eppendorf QuickLock™ Rotor Lid
Rotor Lid
Round Buckets
Universal Buckets
Without Buckets
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Max. Speed Max. RCF Includes Unit Size
15863212 4 x 1,000 mL 4,500 rpm 4,347 x g Universal Buckets Each
15718379 20 × 5 mL 13,100 rpm 20,913 x g Eppendorf QuickLock™ Rotor Lid Each
15758369 4 x 750 mL 4,700 rpm 4,800 x g Round Buckets Each
15768369 4 x 750 mL 4,700 rpm 4,800 x g Without Buckets Each
15633797 48 x 2 mL 13,700 rpm 21,194 x g Eppendorf QuickLock™ Rotor Lid Each
15674614 6 × 50 mL 13,500 rpm 20,130 x g Rotor Lid Each
15873212 4 x 1,000 mL 4,500 rpm 4,347 x g Without Buckets Each
16202411 6 x 250 mL 10,100 rpm 15,054 x g Eppendorf QuickLock™ Rotor Lid Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15863212 Supplier Eppendorf™ Supplier No. 5895200001
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
Eppendorf Rotor for Centrifuge 5910 Ri and 5920 R-precision in refrigerated large-capacity centrifugation. Tailor your experiments with varied capacities and speeds, buckets and adaptors for efficient sample processing. Ideal for high-throughput applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor
Capacity 4 x 1,000 mL
Max. RCF 4,347 x g
Max. Speed 4,500 rpm
For Use With (Equipment) 5910 R Centrifuge
Includes Universal Buckets
No. of Places 4
Model S-4xUniversal