Learn More
Pyrex™ Quickfit™ Rotary Evaporator Splash Head Adapter
Product Code. 12994065
Change view
Click to view available options
Taper:
Cone: 19/26, Socket: 29/32
Cone: 24/29, Socket: 29/32
Cone: 29/32, Socket: 29/32
Unit Size:
Each
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Taper Unit Size
12994065 Cone: 24/29, Socket: 29/32 Each
12904075 Cone: 29/32, Socket: 29/32 Each
15577960 Cone: 19/26, Socket: 29/32 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12994065 Supplier Pyrex™ Supplier No. SH15/24
  • ONLINE EXCLUSIVE PRICE
    137.55 EUR
    107.83 EUR / Pack of 1
    Save 29.72 EUR
    21% Off
View all promotions
Estimated Shipment: 07-12-2026
Protects against foaming products that may seep into a condenser. Pyrex™ Quickfit™ Rotary Evaporator Splash Head Adapter have an anti-climb mechanism.
  • For use with rotary evaporators
  • Protects against liquid ingress into the condenser 
  • Useful for liquids that have tendency to foam 
  • Splash heads feature a 29/32 socket
Specifications
Description Anti-climb
Material Borosilicate Glass
For Use With (Equipment) Rotary evaporators
Quantity 1 Each
Taper Cone: 24/29, Socket: 29/32
Volume (Metric) 150 mL