missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RosetteSep™ HLA Myeloid Cell Enrichment Kit

Product Code. 17167691
Encompass
Change view
Click to view available options
Quantity:
For 20 Tests (10 mL of whole blood per test)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17167691 For 20 Tests (10 mL of whole blood per test) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17167691 Supplier STEMCELL Technologies Supplier No. 15272HLA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunodensity isolation of untouched myeloid cells for lineage-specific chimerism analysis - 20 Tests

  • The RosetteSep™ HLA Myeloid Cell Enrichment Cocktail is designed to enrich myeloid (CD33+) cells from whole blood by negative selection for lineage-specific chimerism analysis.
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD3, CD8, CD19, CD56 and glycophorin A on red blood cells (RBCs).
  • When centrifuged over a buoyant density medium such as RosetteSep™ DM-M (Catalog #15725), the unwanted cells pellet along with the RBCs.
  • The purified myeloid cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium.
  • The desired cells are immediately ready for chimerism analysis or other downstream applications.

Note: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled For Research Use Only. Please note there have been no changes to the form, function and quality assurances associated with these products.

Specifications

Cell Type Myeloid Cells
For Use With (Application) Cell Isolation; In Vitro Diagnostic
Quantity For 20 Tests (10 mL of whole blood per test)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.