Learn More
STEMCELL Technologies RosetteSep™ DM-L Density Medium
Product Code. 17127711
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17127711 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17127711 Supplier STEMCELL Technologies Supplier No. 15705
 Restricted Item
Estimated Shipment: 21-09-2026
Density gradient medium for use with RosetteSep™ CD3+ T cell, CD4+ T cell, CD8+ T cell, and B cell enrichment cocktails - 100 mL
  • RosetteSep™ DM-L is a density gradient medium designed specifically for use with RosetteSep™ cocktails for the enrichment of specific human lymphocyte populations (CD3+ T cells, CD4+ T cells, CD8+ T cells, and B cells) from whole blood. 
  • RosetteSep™ DM-L has a density of 1.081 g/mL. 
Note: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled “For Research Use Only”. Please note there have been no changes to the form, function and quality assurances associated with these products.
Specifications
Quantity 100 mL
Cell Type B Cells, T Cells
For Use With (Application) Cell Isolation; In Vitro Diagnostic